missing translation for 'onlineSavingsMsg'
Learn More

Glycopyrrolate Bromide, TRC

Product Code. 30006633
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30006633 25 mg 25mg
30004544 10 mg 10mg
30004952 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30006633 Supplier Toronto Research Chemicals Supplier No. TRCG65698025MG
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 596-51-0
Molecular Formula C19 H28 N O3 . Br
Molecular Weight (g/mol) 398.33
Synonym Pyrrolidinium, 3-[(2-cyclopentyl-2-hydroxy-2-phenylacetyl)oxy]-1,1-dimethyl-, bromide (1:1), 3-Hydroxy-1,1-dimethylpyrrolidinium bromide α-cyclopentylmandelate (6CI,7CI), Pyrrolidinium, 3-[(cyclopentylhydroxyphenylacetyl)oxy]-1,1-dimethyl-, bromide (9CI), Pyrrolidinium, 3-hydroxy-1,1-dimethyl-, bromide, α-cyclopentylmandelate (8CI), 1,1-Dimethyl-3-hydroxypyrrolidinium bromide α-cyclopentylmandelate, AHR-504, Asecryl, Copyrrolate, Gastrodyn, Glycopyrrolate, Glycopyrrolate bromide, Glycopyrrone bromide, Glycopyrronium bromide, NSC 250836, NSC 251251, NSC 251252, NVA 237, Nodapton, Robanul, Robinul, Seebri, Tarodyl, Tarodyn, β-1-Methyl-3-pyrrolidyl-α-cyclopentylmandelate methobromide
IUPAC Name (1,1-dimethylpyrrolidin-1-ium-3-yl) 2-cyclopentyl-2-hydroxy-2-phenylacetate;bromide
SMILES [Br-].C[N+]1(C)CCC(C1)OC(=O)C(O)(C2CCCC2)c3ccccc3

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C19H28NO3.BrH/c1-20(2)13-12-17(14-20)23-18(21)19(22,16-10-6-7-11-16)15-8-4-3-5-9-15;/h3-5,8-9,16-17,22H,6-7,10-14H2,1-2H3;1H/q+1;/p-1
Formula Weight 397.1253
Percent Purity >95%
Chemical Name or Material Glycopyrrolate Bromide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.