missing translation for 'onlineSavingsMsg'
Learn More

Glycoursodeoxycholic acid, MedChemExpress

Product Code. 16469218
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16469218 10 mg 10mg
16444917 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16469218 Supplier Medchem Express Supplier No. HYN1424
 Restricted Item
Estimated Shipment: 13-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Glycoursodeoxycholic acid, a acyl glycine and a bile acid-glycine conjugate, is a metabolite of ursodeoxycholic acid.

Glycoursodeoxycholic acid, a acyl glycine and a bile acid-glycine conjugate, is a metabolite of ursodeoxycholic acid.

Specifications

Chemical Name or Material Glycoursodeoxycholic acid
CAS 64480-66-6
Color White
Purity Grade Notes Research
Molecular Formula C26H43NO5
Quantity 10 mg
Synonym Ursodeoxycholylglycine
Solubility Information DMSO : 250 mg/mL (556.03 mM; Need ultrasonic)
SMILES O=C(O)CNC(CC[C@@H](C)[C@H]1CC[C@@]2([H])[C@]3([H])[C@@H](O)C[C@]4([H])C[C@H](O)CC[C@]4(C)[C@@]3([H])CC[C@]12C)=O
Molecular Weight (g/mol) 449.62
Formula Weight 449.62
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.