missing translation for 'onlineSavingsMsg'
Learn More

Glycoursodeoxycholic Acid, TRC

Product Code. 30136092
Change view
Click to view available options
Quantity:
50 mg
500 mg
1 g
Unit Size:
1g
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136092 1 g 1g
30134134 500 mg 500mg
30005566 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136092 Supplier Toronto Research Chemicals Supplier No. TRCG6795501G
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64480-66-6
Molecular Formula C26 H43 N O5
Molecular Weight (g/mol) 449.62
Synonym N-[(3α,5β,7β)-3,7-Dihydroxy-24-oxocholan-24-yl]glycine, Glycoursodeoxycholic acid (6CI), N-[(3α,5β,7β)-3,7-Dihydroxy-24-oxocholan-24-yl]glycine, 3α,7β-Dihydroxy-5β-cholanoylglycine, Glycylursodeoxycholic acid, N-(3α,7β-Dihydroxy-5β-cholan-24-oyl)glycine, Ursodeoxycholylglycine
IUPAC Name 2-[[(4R)-4-[(3R,5S,7S,8R,9S,10S,13R,14S,17R)-3,7-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]acetic acid
SMILES C[C@H](CCC(=O)NCC(=O)O)[C@H]1CC[C@H]2[C@@H]3[C@@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3CC[C@]12C

Specifications

Quantity 1 g
InChI Formula InChI=1S/C26H43NO5/c1-15(4-7-22(30)27-14-23(31)32)18-5-6-19-24-20(9-11-26(18,19)3)25(2)10-8-17(28)12-16(25)13-21(24)29/h15-21,24,28-29H,4-14H2,1-3H3,(H,27,30)(H,31,32)/t15-,16+,17-,18-,19+,20+,21+,24+,25+,26-/m1/s1
Formula Weight 449.3141
Chemical Name or Material Glycoursodeoxycholic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.