missing translation for 'onlineSavingsMsg'
Learn More

Glycyl-DL-phenylalanine, TRC

Product Code. 30179364
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30179364 5 g 5g
30179560 500 mg 500mg
30179262 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179364 Supplier Toronto Research Chemicals Supplier No. TRCG1153305G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3321-03-7
Molecular Formula C11H14N2O3
Molecular Weight (g/mol) 222.24
Synonym (2S)-2-(2-Aminoacetamido)-3-phenylpropanoic Acid, (2S)-2-[(2-Azaniumylacetyl)amino]-3-phenylpropanoate;(S)-2-(2-Aminoacetamido)-3-phenylpropanoicacid;43: PN: US20130123467 SEQID: 48 claimed protein, 7: PN: WO2014063098 TABLE: 5 claimed protein, 85: PN: EP2161028 PAGE: 10 claimed protein, Glycylphenylalanine, N-Glycyl-L-phenylalanine, N-Glycylphenylalanine, NSC 88881;
IUPAC Name (2S)-2-[(2-aminoacetyl)amino]-3-phenylpropanoic acid
SMILES NCC(=O)N[C@@H](Cc1ccccc1)C(=O)O

Specifications

InChI Formula InChI=1S/C11H14N2O3/c12-7-10(14)13-9(11(15)16)6-8-4-2-1-3-5-8/h1-5,9H,6-7,12H2,(H,13,14)(H,15,16)/t9-/m0/s1
Quantity 5 g
Formula Weight 222.1004
Shipping Condition Room Temperature
Chemical Name or Material Glycyl-DL-phenylalanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.