missing translation for 'onlineSavingsMsg'
Learn More

Glyoxal (5% in 250mL in H2O), TRC

Product Code. 17980625
Change view
Click to view available options
Quantity:
25 mL
250 mL
Unit Size:
250mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17980625 250 mL 250mL
17911235 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17980625 Supplier Toronto Research Chemicals Supplier No. TRCG754570250ML
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-22-2
Molecular Formula C2H2O2
Molecular Weight (g/mol) 58.04
Synonym Ethanedial, Glyoxal (8CI), 1,2-Ethanedione, Aurarez 136, Biformal, Biformyl, Cartabond GH, Cartabond GHF, Daicel GY 60, Diformyl, Earth Works Linkup Plus, Ethanedione, Freechem 40DL, GX, GX (aldehyde), Glyfix CS 50, Glyoxal 40L, Glyoxal T 40, Glyoxal aldehyde, Glyoxazal, Glyoxazal GX, Glyoxylaldehyde, Gohsezal P, Oxal, Oxalaldehyde, Permafresh 114, Protorez BLF-C, XH 536
IUPAC Name oxaldehyde
SMILES O=CC=O

Specifications

Quantity 250 mL
InChI Formula InChI=1S/C2H2O2/c3-1-2-4/h1-2H
Formula Weight 58.0055
Concentration or Composition (by Analyte or Components) Glyoxal 5%
Chemical Name or Material Glyoxal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.