missing translation for 'onlineSavingsMsg'
Learn More

Glyoxal-bis(2-hydroxyanil), ≥97.0% (from N), Solstice

Product Code. 15683940 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
25 g
6 x 25 g
Packaging:
Glass Bottle
Unit Size:
25g
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15683940 6 x 25 g Glass Bottle Pack of 6
15673940 25 g Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15683940 Supplier Honeywell-Fluka Supplier No. 332556X25G
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Indicator for metal titration

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1149-16-2
Molecular Formula C14H12N2O2
Molecular Weight (g/mol) 240.26
MDL Number MFCD00002183
InChI Key OMJNMRJMNUREOJ-KOBPNRPCSA-N
PubChem CID 5356911
SMILES OC1=CC=CC=C1N\C=C\N=C1C=CC=CC1=O

Specifications

Melting Point 203°C to 211°C
UN Number NONH for all modes of transport
Beilstein 2808815
IUPAC Name 6-{[(1E)-2-[(2-hydroxyphenyl)amino]ethenyl]imino}cyclohexa-2,4-dien-1-one
Formula Weight 240.26g/mol
Percent Purity ≥97% (from N)
Odor Odourless
Packaging Glass Bottle
Suitability Passes test for suitable for metal definition
Color Yellow
Quantity 6 x 25 g
Chemical Name or Material Glyoxal-bis(2-hydroxyanil)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.