missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Glyoxal, pure, 40 wt.% solution in water

Product Code. 10582551 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
25 L
Packaging:
Glass bottle
Metal Drum
Unit Size:
1 L
2.5 L
25 L
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10582551 500 mL Glass bottle 500mL
10081700 1 L Glass bottle 1 L
10091700 2.5 L Glass bottle 2.5 L
10593314 25 L Metal Drum 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10582551 Supplier Thermo Scientific Chemicals Supplier No. 156225000
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 107-22-2 | C2H2O2 | 58.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Glyoxal
Name Note 40 wt.% Solution in Water
CAS 107-22-2, 7732-18-5
Melting Point -14.0°C
Color Colorless to Yellow
Density 1.2650g/mL
Boiling Point 104.0°C
Flash Point >104°C
Packaging Glass bottle
Molecular Formula C2H2O2
Linear Formula HCOCHO
MDL Number MFCD00006957
Quantity 500 mL
Beilstein 01, 759
Fieser 01,413
Merck Index 15, 4544
Specific Gravity 1.265
Synonym glyoxal, ethanedial, oxalaldehyde, 1,2-ethanedione, glyoxylaldehyde, biformal, biformyl, diformal, diformyl, aerotex glyoxal 40
Solubility Information Solubility in water: miscible.
InChI Key LEQAOMBKQFMDFZ-UHFFFAOYSA-N
SMILES C(=O)C=O
IUPAC Name oxaldehyde
Molecular Weight (g/mol) 58.04
PubChem CID 7860
ChEBI CHEBI:34779
Viscosity 8 mPa.s (20°C)
Formula Weight 58.04
Percent Purity 39 to 41% (Titrimetry other)
Grade Pure
Physical Form Liquid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Glyoxal, 40 wt% solution in water
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H341-Suspected of causing genetic defects.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.