missing translation for 'onlineSavingsMsg'
Learn More

Glyoxime, 98+%, moistened with ca 20% water

Product Code. 11331837
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11331837 10 g 10g
11341837 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11331837 Supplier Thermo Scientific Alfa Aesar Supplier No. L05130.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 557-30-2 | C2H4N2O2 | 88.066 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 557-30-2
Molecular Formula C2H4N2O2
Molecular Weight (g/mol) 88.066
MDL Number MFCD00013942
InChI Key RUFIRPVAEJIIIS-OWOJBTEDSA-N
Synonym glyoxime, ethanedial, dioxime, ethanedione dioxime, pik-off, glyoxaldioxime, glyoxal dioxime, ethanedial dioxime, glyoxal-dioxime, dioxime ethanedial, glyoxal, dioxime
PubChem CID 5354753
IUPAC Name N-[(E)-2-nitrosoethenyl]hydroxylamine
SMILES C(=CN=O)NO

Specifications

Melting Point ∽169°C (decomposition)
Quantity 10 g
UN Number UN2926
Beilstein 1699539
Formula Weight 88.07
Percent Purity ≥98%
Assay moistened with ≈20% water
Chemical Name or Material Glyoxime

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.