missing translation for 'onlineSavingsMsg'
Learn More

Glyoxime, 98+%, moistened with ca 20% water

Product Code. 11341837
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11341837 50 g 50g
11331837 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11341837 Supplier Thermo Scientific Alfa Aesar Supplier No. L05130.18
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 557-30-2 | C2H4N2O2 | 88.066 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 557-30-2
Molecular Formula C2H4N2O2
Molecular Weight (g/mol) 88.066
MDL Number MFCD00013942
InChI Key RUFIRPVAEJIIIS-OWOJBTEDSA-N
Synonym glyoxime, ethanedial, dioxime, ethanedione dioxime, pik-off, glyoxaldioxime, glyoxal dioxime, ethanedial dioxime, glyoxal-dioxime, dioxime ethanedial, glyoxal, dioxime
PubChem CID 5354753
IUPAC Name N-[(E)-2-nitrosoethenyl]hydroxylamine
SMILES C(=CN=O)NO

Especificaciones

Melting Point ∽169°C (decomposition)
Quantity 50 g
UN Number UN2926
Beilstein 1699539
Formula Weight 88.07
Percent Purity ≥98%
Assay moistened with ≈20% water
Chemical Name or Material Glyoxime

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.