missing translation for 'onlineSavingsMsg'
Learn More

Glyoxylic acid monohydrate, 97%

Product Code. 11473067
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11473067 25 g 25g
11483067 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11473067 Supplier Thermo Scientific Alfa Aesar Supplier No. A16058.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 563-96-2 | C2H4O4 | 92.05 g/mol

Glyoxylic acid is a reagent for sulfinylmaleate synthesis. It is one of the chemicals used in the Hopkins Cole reaction. It is involved to check the presence of tryptophan in proteins. It is condensed with urea and 1,2-diaminobenzene to form heterocycles. It also undergoes Friedel-Crafts and cyclocondensation reactions to form bis(pentamethylphenyl) acetic acid and a beta-carboline respectively. It is employed in the synthesis of a sulfinylmaleate, which serves as an efficient dienophile for enantioselective Diels-Alder cycloadditions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glyoxylic acid is a reagent for sulfinylmaleate synthesis. It is one of the chemicals used in the Hopkins Cole reaction. It is involved to check the presence of tryptophan in proteins. It is condensed with urea and 1,2-diaminobenzene to form heterocycles. It also undergoes Friedel-Crafts and cyclocondensation reactions to form bis(pentamethylphenyl) acetic acid and a beta-carboline respectively. It is employed in the synthesis of a sulfinylmaleate, which serves as an efficient dienophile for enantioselective Diels-Alder cycloadditions.

Solubility
Soluble in water, dimethyl sulfoxide and alcohol. Insoluble in diethyl ether and hydrocarbons.

Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible withbases, oxidizing agents and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 563-96-2
Molecular Formula C2H4O4
Molecular Weight (g/mol) 92.05
MDL Number MFCD00149497
InChI Key MOOYVEVEDVVKGD-UHFFFAOYSA-N
Synonym glyoxylic acid monohydrate, glyoxylic acid hydrate, 2-oxoacetic acid hydrate, oxaldehydic acid hydrate, glyoxalic acid monohydrate, glyoxalate, glyoxylate hydrate, glyoxalic acid hydrate, oxoacetate monohydrate, glyoxylic acid-hydrate, oxoacetic acid hydrate
PubChem CID 15620607
IUPAC Name oxaldehydic acid;hydrate
SMILES C(=O)C(=O)O.O

Specifications

Melting Point 48°C to 52°C
Boiling Point 100°C
Flash Point >110°C (230°F)
Quantity 25 g
UN Number UN3261
Beilstein 969535
Sensitivity Hygroscopic
Merck Index 14,4511
Solubility Information Soluble in water,dimethyl sulfoxide and alcohol. Insoluble in diethyl ether and hydrocarbons.
Formula Weight 92.06 (74.04 Anhydrous)
Percent Purity 97%
Chemical Name or Material Glyoxylic acid monohydrate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Glyoxylic acid monohydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Glyoxylic acid monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.