missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glypican 2 Rat anti-Mouse, Clone: 300712, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB23551-SP
This item is not returnable.
View return policy
Description
Glypican 2 Monoclonal specifically detects Glypican 2 in Mouse samples. It is validated for Western Blot.Specifications
| Glypican 2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q8BKV1 | |
| Gpc2 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Glypican 2 Lys27-Leu554 Accession # Q8BKV1 | |
| 25 μg | |
| Primary | |
| Detects mouse Glypican 2 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human Glypican 2, 3, 6, or recombinant mouse Glypican 5 is observed. | |
| Mouse | |
| Purified |
| Western Blot | |
| 300712 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 2410016G05Rik, Cerebroglycan, GPC2 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 221914 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction