missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals GM 6001

Product Code. 15457039
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
This item is not returnable. View return policy

Product Code. 15457039

Brand: Thermo Scientific Chemicals J65687.M^

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

GM 6001 is used to inhibits a wide variety of matrix metalloproteases with Kis in the nanomolar range. Active in a number of animal models of diseases where matrix metalloproteases are thought to be involved.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142880-36-2
Molecular Formula C20H28N4O4
Synonym Galardin; N-[(2R)-2-(Hydroxamidocarbonylmethyl)-4-methylpentanoyl]-L-tryptophan methylamide

Specifications

Quantity 1 mg
Description Off-white
Content And Storage 2°C to 8°C
For Use With (Application) A potent, cell-permeable, broad-spectrum hydroxamic acid inhibitor of matrix metalloproteinases
Formula Weight 388.50
Sensitivity Moisture Sensitive
Product Type GM 6001
Form Solid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.