missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ GM 6001

Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
Broad spectrum MMP inhibitor. Reduces infarct volume following middle cerebral artery occlusion in an ischemic mouse model. Also inhibits human skin fibroblast collagenase (Ki = 0.4 nM).
Specifications
Specifications
| CAS | 142880-36-2 |
| Quantity | 10 mg |
| Molecular Formula | C20H28N4O4 |
| Formula Weight | 388.46 |
| Purity | >95% |
| Molecular Weight (g/mol) | 388.468 |
| Chemical Name or Material | GM 6001 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction