missing translation for 'onlineSavingsMsg'
Learn More

AcroBiosystems GMP ActiveMax™ Human T cell Activation/Expansion CD3/CD28 Beads

Product Code. 31453495 Shop All ACROBiosystems Products
Change view
Click to view available options
Quantity:
400 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31453495 400 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31453495 Supplier AcroBiosystems Supplier No. GMPMBS001400mg
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

GMP ActiveMax™ Human T cell Activation/Expansion CD3/CD28 Beads is expressed from Human

GMP ActiveMax™ Human T cell Activation/Expansion CD3/CD28 Beads is expressed from Human
TRUSTED_SUSTAINABILITY

Specifications

Formulation GMP ActiveMax Human T cell Activation/Expansion CD3/CD28 Beads is supplied with 5x107 beads/mL in PBS, pH 7.4, with 0.1% recombinant human serum albumin (recombinant HSA).
Content And Storage The product MUST be stored at 2°C to 8°C upon receipt; This product is stable after storage at 2°C to 8°C for 5 years under sterile conditions.
Particle Size 5.5 μm
Quantity 400 mg
For Use With (Application) T cell Activation/Expansion
Species Human
Sufficient For 4 x 109 beads
Regulatory Status RUO
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.