missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Bone Morphogenetic Protein 4 (BMP4) is a member of growth factor of the TGF-beta superfamily that plays essential roles in many developmental processes, including neurogenesis, vascular development, angiogenesis and osteogenesis. BMP-4 Initiates the canonical BMP signaling cascade by associating with type I receptor BMPR1A and type II receptor BMPR2. BMP-4 can acts in concert with PTHLH/PTHRP to stimulate ductal outgrowth during embryonic mammary development and to inhibit hair follicle induction.
Spécification
Spécification
| Accession Number | Uniprot |
| Characterization | BMP-4 Structure This protein carries no tag. The protein has a calculated MW of 13.1 kDa. The protein migrates as 21 kDa±3 kDa when calibrated against under reducing (R) condition (SDS-PAGE) due to glycosylation. |
| Format | Powder-GMP |
| Formulation | Lyophilized from 0.22 μm filtered solution in 20 mM Citric acid, pH 2.2 with protectants. Contact us for customized product form or formulation. |
| Quality Control Testing | <0.5 ng/μg of protein tested by ELISA.∣<0.02 ng/μg of protein tested by qPCR.∣Less than 10 EU/mg, tested by the LAL method in compliance with USP 85 and Ph. Eur. 2.6.14.∣Sterility testing was performed using the membrane filtration method in compliance with USP &CM and Ph. Eur. 2.6.1.∣Negative |
| Quantity | 10 mg |
| Source | GMP Human BMP-4 Protein (GMP-BM4H36) is expressed from CHO cells. It contains AA Ser 293 - Arg 408 (Accession # ). Predicted N-terminus: Ser 293 Request for sequence |
| Biological Activity | GMP Human BMP-4 Protein (Cat. No. GMP-BM4H36) stimulates Human BMP (Luc) HEK293 Reporter cells. The typical EC50 for this effect is 2.563 ng/mL (QC tested).∣GMP Human BMP-4 Protein (Cat. No. GMP-BM4H36) could promote the differentiation of iPSCs into hematopoietic stem and progenitor cells in a feeder-free system at Day 14, with high expression of CD34+ CD45+. GMP Human BMP-4 Protein (Cat. No. GMP-BM4H36) holds a similar performance with Human BMP-4 Protein, premium grade (Cat. No. BM4-H5317).∣The Cell based assay shows that GMP Human BMP-4 Protein (Cat. No. GMP-BM4H36) is stable at 37°C for 7 hours.∣The Cell based assay shows that GMP Human BMP-4 Protein (Cat. No. GMP-BM4H36) is stable at 4°C for 180 days.∣The Cell based assay shows that GMP Human BMP-4 Protein (Cat. No. GMP-BM4H36) is stable after freezing and thawing 3 times. |
| Species | Human |
| Sterility | Sterile |
| Afficher plus |
Nom du produit
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Vous avez repéré une opportunité d'amélioration ?