missing translation for 'onlineSavingsMsg'
Learn More

AcroBiosystems GMP Human IL-1 beta Protein

Product Code. 31451805 Shop All ACROBiosystems Products
Change view
Click to view available options
Quantity:
10 mg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31451805 50 μg
31453799 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 31451805 Supplier AcroBiosystems Supplier No. GMPILBH1650ug
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Recombinant Protein

Interleukin-1 beta (IL-1β) is a non-secreted proinflammatory cytokine produced mainly by activated macrophages, as well as neutrophils, epithelial cells, and endothelial cells. It possesses metabolic, physiological, haematopoietic activities, and plays one of the central roles in the regulation of the immune responses. Both IL-1α and IL-1β binds to the same receptor and has similar but not identical biological properties; The mature human IL1β shares 96 % amino acid sequence identity with rhesus and 67 % -78 % with canine, mouse and rat IL-1β.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Uniprot
Characterization IL-1 beta Structure This protein carries no tag. The protein has a calculated MW of 17.5 kDa. The protein migrates as 18 kDa±3 kDa under reducing (R) condition (SDS-PAGE).
Format Powder-GMP
Formulation Lyophilized from 0.22 μm filtered solution in PBS, pH 7.4 with protectants. Contact us for customized product form or formulation.
Quality Control Testing <0.5 ng/μg of protein tested by ELISA.∣<0.02 ng/μg of protein tested by qPCR.∣Less than 10 EU/mg, tested by the LAL method in compliance with USP 85 and Ph. Eur. 2.6.14.∣Sterility testing was performed using the membrane filtration method in compliance with USP &CM and Ph. Eur. 2.6.1.∣Negative
Quantity 50 μg
Source GMP Human IL-1 beta Protein (GMP-ILBH16) is expressed from E. coli cells. It contains AA Ala 117 - Ser 269 (Accession # ). Request for sequence
Biological Activity GMP Human IL-1 beta Protein (Cat. No. GMP-ILBH16) stimulates proliferation of D10.G4.1 cells. The specific activity of GMP Human IL-1 beta Protein is>5.00 x 10^7 IU/mg, which is calibrated against WHO International Standard INTERLEUKIN-1 BETA (IL-1ß, Human rDNA derived) (NIBSC code: 86/680) (QC tested).∣The Cell based assay shows batch-to-batch consistency between Acro's GMP and PG IL-1beta.∣MALDI-TOF analysis of GMP Human IL-1 beta Protein (Cat. No. GMP-ILBH16). The labeled peaks at 17372.1 Da and 17301.3 Da correspond to the calculated molecular masses, 17377 Da (aa 117-269) and 17306 Da (aa 118-269), respectively (Routinely tested).
Species Human
Sterility Sterile
Content And Storage -20°C to -70°C for 5 years in lyophilized state; -70°C for 12 months under sterile conditions after reconstitution. Upon receipt, store it immediately at -20°C or lower for long term storage.
Endotoxin Level Less than 10 EU/mg, tested by the LAL method in compliance with USP 85 and Ph. Eur. 2.6.14.
Shipping Condition This product is supplied and shipped with blue ice, please inquire the shipping cost.
Purity or Quality Grade 0.95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.