missing translation for 'onlineSavingsMsg'
Learn More

GeneBLAzer™ GeneBLAzer™ GnRHR-NFAT-bla CHO-K1 Cells

Product Code. 10276934
Change view
Click to view available options
Quantity:
1 vial
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10276934 1 vial Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10276934 Supplier GeneBLAzer™ Supplier No. K1854

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

GeneBLAzer™ GnRHR-NFAT-bla CHO-K1 Cells contain the human gonadotropin-releasing hormone receptor.

  • Accession Number: NM_000406.2
  • Population Status: Single Cell Clone Origin
  • GeneBLAzer™ GnRHR-NFAT-bla CHO-K1 cells contain the human gonadotropin-releasing hormone receptor (GnRHR), (Accession # NM_000406.2) stably integrated into the CellSensor™ NFAT-bla CHO-K1 cell line. CellSensor™ NFAT-bla CHO-K1 (Cat No. K1854A) cells contain a beta-lactamase reporter gene under control of the NFAT Response Element. GeneBLAzer™ GnRHR-NFAT-bla CHO-K1 cells are functionally validated for Z’-factor and EC50 concentrations of Luteinizing Hormone-Releasing Hormone (LH-RH). In addition, GeneBLAzer™ GnRHR-NFAT-bla CHO-K1 cells have been tested for assay performance under variable conditions.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NM_000406.2
Assay Entry Cell-based beta lactamase reporter gene
Cell State Dividing Cells
Content And Storage GeneBLAzer™ GnRHR-NFAT-bla CHO-K1 cells are shipped on dry ice, 8 x 10^6 cells⁄vial. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use.
Detection Method Fluorescence
For Use With (Application) FRET
Human Protein Class G-Protein Coupled Receptor (GPCR)
Population Status Single Cell Clone Origin
Product Line GeneBLAzer
Quantity 1 vial
Readout End Point
Target Entry GnRHR
Cell Line CHO-K1
System Type GeneBLAzer™
Druggable Target GPCR
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.