missing translation for 'onlineSavingsMsg'
Learn More

Go 6983, MedChemExpress

Product Code. 16478477
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16478477 5 mg 5mg
16441967 50 mg 50mg
16416286 100 mg 100mg
16405818 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16478477 Supplier Medchem Express Supplier No. HY13689
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Go 6983 is a pan-PKC inhibitor against for PKCα, PKCβ, PKCγ, PKCδ and PKCζ with IC50 of 7 nM, 7 nM, 6 nM, 10 nM and 60 nM, respectively.

Go 6983 is a pan-PKC inhibitor against for PKCα, PKCβ, PKCγ, PKCδ and PKCζ with IC50 of 7 nM, 7 nM, 6 nM, 10 nM and 60 nM, respectively.

Specifications

Chemical Name or Material Go 6983
CAS 133053-19-7
Color Red
Purity Grade Notes Research
Molecular Formula C26H26N4O3
Quantity 5 mg
Synonym Gö 6983 Goe 6983
Solubility Information DMSO : ≥ 34 mg/mL (76.83 mM)
SMILES O=C(C(C1=CN(CCCN(C)C)C2=C1C=C(OC)C=C2)=C3C4=CNC5=C4C=CC=C5)NC3=O
Molecular Weight (g/mol) 442.51
Formula Weight 442.51
Percent Purity 95.61%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.