missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Goat anti-Bovine IgG (H+L) Secondary Antibody, Agarose
Goat Polyclonal Secondary Antibody
Brand: Invitrogen™ PA130750
This item is not returnable.
View return policy
Description
PA1-30750 detects Bovine IgG from bovine samples.
Anti-Bovine secondary antibodies are affinity-purified antibodies with well-characterized specificity for bovine (cow, ox) immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
| Bovine IgG (H+L) | |
| Polyclonal | |
| Agarose | |
| Goat | |
| IgG fraction | |
| RUO | |
| Bovine | |
| Secondary Antibody | |
| IgG |
| Affinity Purification | |
| 9.2 mg/mL | |
| PBS with 0.01% sodium azide; pH 7.2 | |
| Bovine IgG whole molecule. | |
| 10 mg | |
| Secondary | |
| 4°C, do not freeze | |
| Slurry |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction