Learn More
Invitrogen™ Goat anti-Chicken IgY (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor™ Plus 405
Descripción
To minimize cross-reactivity, the goat anti-chicken IgY whole antibodies have been affinity-purified and cross-adsorbed against chicken serum containing non-immunoglobulin chicken serum proteins. Cross-adsorption or pre-adsorption is a purification step to increase specificity of the antibody resulting in higher sensitivity and less background staining. The secondary antibody solution is passed through a column matrix containing immobilized serum proteins from potentially cross-reactive species. Only the nonspecific-binding secondary antibodies are captured in the column, and the highly specific secondaries flow through. The benefits of this extra step are apparent in multiplexing/multicolor-staining experiments (e.g., flow cytometry) where there is potential cross-reactivity with other primary antibodies or in tissue/cell fluorescent staining experiments where there may be the presence of endogenous immunoglobulins. Using conjugate solutions: Centrifuge the protein conjugate solution briefly in a microcentrifuge before use; add only the supernatant to the experiment. This step will help eliminate any protein aggregates that may have formed during storage, thereby reducing nonspecific background staining. Because staining protocols vary with application, the appropriate dilution of antibody should be determined empirically. Specificity: This antibody binds to heavy chains on chicken IgY and light chains on all chicken immunoglobulins. This antibody does not bind non-immuno...
Especificaciones
Especificaciones
| Antigen | Chicken IgY (H+L) Cross-Adsorbed |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 2 mg/mL |
| Conjugate | Alexa Fluor Plus 405 |
| Formulation | proprietary buffer with 0.016% Bromonitrodioxane, 0.016% Methylisothiazolone; pH 6.5 |
| Host Species | Goat |
| Immunogen | Purified Chicken IgY, Heavy and Light chains. |
| Purification Method | Affinity chromatography |
| Quantity | 1 mg |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.