Learn More
Invitrogen™ Goat anti-Human IgA (Heavy chain) Recombinant Secondary Antibody
Description
This Goat anti-Human IgA (Heavy chain), Recombinant Monoclonal Antibody specifically detects human IgA heavy chain with no cross-reactivity to human IgG or IgM or various species immunoglobulins. The sensitivity and specificity of each lot is confirmed by ELISA. Recombinant antibodies are produced using specific genes that code for the desired antibodies. These genes are cloned into an expression vector and expressed in vitro. The advantages of recombinant antibodies include: better specificity, animal origin-free formulation, and lot-to-lot consistency. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
Specifications
Specifications
| Antigen | Human IgA (Heavy chain) |
| Applications | Peptide Array, ELISA, Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | 43H46L44 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Host Species | Goat |
| Immunogen | Purified Human IgA. |
| Purification Method | Protein G |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.