missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Human IgA/IgG/IgM (H+L) Secondary Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-69264
This item is not returnable.
View return policy
Description
IgA/IgG/IgM (H+L) Polyclonal antibody specifically detects IgA/IgG/IgM (H+L) in Human samples. It is validated for Immunoprecipitation
Specifications
| IgA/IgG/IgM (H+L) | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from PBS (pH 7.2) | |
| Highly purified IgG and pools of homogenous IgA and IgM isolated from human serum. Freund's complete adjuvant is used in the first step of the immunization procedure. | |
| 10 mg | |
| Secondary | |
| Human | |
| Lyophilized |
| Immunoprecipitation | |
| LYOPH | |
| Immunoprecipitation | |
| Goat | |
| Ion exchange chromatography | |
| RUO | |
| Reconstitute by by adding 1 mL sterile distilled water, spun down to remove insoluble particles, divided into small aliquots, frozen and stored at or below -20C. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction