Learn More
Goat anti-Human IgD Heavy Chain Secondary Antibody, Janelia Fluor 646, Novus Biologicals™
Goat Polyclonal Antibody
Brand:  Novus Biologicals NBP2-68528JF646
Product Code. 30228348
  • 582.00 € / 1mL
Description
IgD Heavy Chain Polyclonal antibody specifically detects IgD Heavy Chain in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IgD Heavy Chain
Polyclonal
50mM Sodium Borate
Goat
Affinity purified
RUO
Human
Purified
ELISA
Janelia Fluor 646
Constant Region of Heavy Chain of IgD, Ig Delta Chain C Region, Ig Delta Chain C Region NIG-65, Ig Delta Chain C Region WAH, Immunoglobulin Delta, immunoglobulin heavy constant delta
Human IgD paraproteins
1 mL
Secondary
Store at 4°C in the dark.