missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Human IgG Fc Secondary Antibody, DyLight 550, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-75816
158.95 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
IgG Fc Polyclonal antibody specifically detects IgG Fc in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunomicroscopy.
Especificaciones
| IgG Fc | |
| Polyclonal | |
| DyLight 550 | |
| O75015, O75015 | |
| Purified Human IgG, Fc fragment | |
| 1 mg | |
| Secondary | |
| Rehydrate with 1.1 ml of deionized water and let stand 30 minutes at room temperature to dissolve. (Product has been overfilled to ensure complete recovery.) Centrifuge to remove any particulates. Prepare fresh working dilution daily. | |
| Store lyophilized antibody at 4°C. Mix reconstituted liquid with an equal volume of glycerol, aliquot contents and freeze at -20°C or below for long term storage. |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunomicroscopy | |
| LYOPH | |
| Flow Cytometry 1:20 - 1:2000, Immunocytochemistry/Immunofluorescence, Immunomicroscopy 1:20 - 1:2000, Immunocytochemistry | |
| Goat | |
| Affinity purified | |
| RUO | |
| Based on IEP, this antibody reacts with heavy gamma chains on human IgG | |
| Human | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido