missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Human IgG/IgM (H+L) Secondary Antibody, HRP (Horseradish Peroxidase), Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-66687
This item is not returnable.
View return policy
Description
IgG/IgM (H+L) Polyclonal specifically detects IgG/IgM (H+L) in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry.
Specifications
| IgG/IgM(H+L) | |
| Polyclonal | |
| HRP | |
| PBS, 0.2% BSA with 0.05% Pro-Clean 400 | |
| Human IgG+IgM-heavy and light chain | |
| 1 mL | |
| Human | |
| IgG |
| ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot | |
| 1mg/mL | |
| Western Blot 1:5000 - 1:50000, ELISA 1:10000 - 1:00000, Immunohistochemistry 1:200 - 1:5000, Immunocytochemistry 1:200 - 1:5000 | |
| Goat | |
| Affinity Purified | |
| Secondary | |
| Store at 4°C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction