Learn More
Invitrogen™ Goat anti-Human IgG (Kappa light chain) Recombinant Secondary Antibody
Goat Recombinant Monoclonal Secondary Antibody
113.00 € - 274.00 €
Specifications
| Antigen | Human IgG (Kappa light chain) |
|---|---|
| Clone | 81H59L2 |
| Concentration | 1.0 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17835702
|
Invitrogen™
A56854 |
250 μL |
274.00 €
250µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17846502
|
Invitrogen™
A5685450UL |
50 μL |
113.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Goat-anti-Human IgG (Kappa Light Chain), Recombinant Secondary Antibody specifically detects human IgG Kappa Light chain with no cross-reactivity to Lambda light chain, heavy chain, and other species immunoglobulins. Recombinant antibodies are produced using specific genes that code for the desired antibodies. These genes are cloned into an expression vector and expressed in vitro. The advantages of recombinant antibodies include better specificity, animal origin-free formulation, and lot-to-lot consistency. The sensitivity and specificity of each lot are confirmed using ELISA. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Specifications
| Human IgG (Kappa light chain) | |
| 1.0 mg/mL | |
| ELISA, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Goat | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Secondary | |
| Protein G |
| 81H59L2 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human | |
| Purified Human IgG. | |
| Secondary Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.