missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Goat anti-Human IgM Fc Secondary Antibody
Goat Polyclonal Secondary Antibody
Marca: Invitrogen™ H15000
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Goat anti-Human IgM Fc Secondary Antibody for Flow
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifica
| Human IgM Fc, Goat anti-Human IgM Fc Secondary Antibody | |
| Polyclonal | |
| PBS with 0.1% sodium azide | |
| Purified | |
| ASR | |
| Human | |
| Secondary Antibody | |
| IgG |
| Flow Cytometry | |
| Unconjugated | |
| Goat | |
| 2 mL | |
| Secondary | |
| 4°C, store in dark | |
| Liquid |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto