Learn More
Goat anti-Mouse Ig, Polyclonal, Secondary Antibody, BD
Brand: BD Biosciences 554002
Code : NEW
Additional Details : Weight : 0.00100kg
Description
X
Host Species: Goat
Species Reactivity: Mouse
ELISA, Western Blotting
Specifications
Goat anti-Mouse Ig | |
POLYCLONAL | |
Goat | |
RUO | |
Secondary | |
Ig |
Polyclonal | |
HRP | |
1 mL | |
Immunology | |
Mouse |
Safety and Handling
- HRP Goat Anti-Mouse Ig
Signal Word
- Warning
Hazard Category
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : CMIT/MIT mixture (3:1) - a mixture of: 5-chloro-2-methyl-4-isothiazolin-3-one ¢EC No 247-500-7! and