Learn More
Invitrogen™ Goat anti-Mouse IgA Secondary Antibody
Goat Polyclonal Secondary Antibody
Brand:  Invitrogen™ PA129741
Product Code. 11526792
  • 239.00 € / 500µg
Description
Goats were immunized with highly purified mouse serum IgA in Freunds adjuvant to produce PA1-29741.
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Mouse IgA
Polyclonal
Unconjugated
Goat
Antigen affinity chromatography
RUO
Mouse
Secondary Antibody
IgG
ELISA, Immunoprecipitation, Western Blot, Western Blot
1 mg/mL
PBS with 0.1% sodium azide; pH 7.2
Goats were immunized with highly purified mouse serum IgA in Freunds adjuvant.
500 μg
Secondary
4°C