Learn More
Goat anti-Mouse IgG Fc Secondary Antibody, DyLight 650, Novus Biologicals™
Goat Polyclonal Antibody
Brand:  Novus Biologicals NBP1-76006
158.95 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
Product Code. 18250746
  • 204.00 € / 1mg
Description
IgG Fc Polyclonal antibody specifically detects IgG Fc in Mouse samples. It is validated for Flow Cytometry, Immunomicroscopy.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IgG Fc
Polyclonal
DyLight 650
Goat
Affinity purified
RUO
Based on IEP, this antibody reacts with heavy gamma chains on mouse IgG
Mouse
IgG
Flow Cytometry, Immunomicroscopy
LYOPH
Flow Cytometry 1:20 - 1:2000, Immunomicroscopy 1:20 - 1:2000
Purified Mouse IgG, Fc fragment
1 mg
Secondary
Rehydrate with 1.1 ml of deionized water and let stand 30 minutes at room temperature to dissolve. (Product has been overfilled to ensure complete recovery.) Centrifuge to remove any particulates. Prepare fresh working dilution daily.
Store lyophilized antibody at 4°C. Mix reconstituted liquid with an equal volume of glycerol, aliquot contents and freeze at -20°C or below for long term storage.