Learn More
Invitrogen™ Goat anti-Mouse IgG (H+L), Superclonal™ Recombinant Secondary Antibody, Alexa Fluor™ Plus 488, Invitrogen™
Goat Recombinant Polyclonal Secondary Antibody
121.00 € - 361.00 €
Specifications
| Antigen | Mouse IgG (H+L) |
|---|---|
| Concentration | 1 mg/mL |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Superclonal |
| Conjugate | Alexa Fluor Plus 488 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17619306
|
Invitrogen™
A5505850UL |
50 μL |
121.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17609306
|
Invitrogen™
A55058 |
250 μL |
361.00 €
250µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Recombinant antibodies are produced using specific genes that code for the desired antibodies. These genes are cloned into an expression vector and expressed in vitro. The advantages of recombinant antibodies include: better specificity and superior lot-to-lot consistency. Minimal cross-reactivity with rabbit, rat, human, bovine, guinea pig, and donkey IgG is observed. Obtain superior images with the new Invitrogen Alexa Fluor Plus secondary antibodies. Make your low abundant targets visible, minimize time spent optimizing, and make your precious samples count. Alexa Fluor Plus secondary antibodies provide brighter signal compared to leading Alexa Fluor secondary antibodies, providing you with better signal-to-noise for your critical experiments. Alexa Fluor Plus secondary antibodies represent an advancement in fluorescent conjugate technology. Alexa Fluor Plus secondary antibodies are conjugated using new, proprietary dye chemistry so you can generate stunning images. Using conjugate solutions: Centrifuge the protein conjugate solution briefly in a microcentrifuge before use; add only the supernatant to the experiment. This step will help eliminate any protein aggregates that may have formed during storage, thereby reducing nonspecific background staining. Because staining protocols vary with application, the appropriate dilution of antibody should be determined empirically.
Mouse IgG (H+L) Recombinant Polyclonal antibody specifically detects Mouse IgG (H+L) in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot.Specifications
| Mouse IgG (H+L) | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Alexa Fluor Plus 488 | |
| Goat | |
| Mouse | |
| Purified Mouse Immunoglobulin. | |
| Secondary | |
| 4°C, store in dark, DO NOT FREEZE! |
| 1 mg/mL | |
| Recombinant Superclonal | |
| Liquid | |
| RUO | |
| Proprietary buffer with 0.008% Bromonitrodioxane, 0.008% Methylisothiazolone; pH 6.5 | |
| IgG | |
| Affinity chromatography | |
| Secondary Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.