missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Goat anti-Mouse IgG1 Cross-Adsorbed Secondary Antibody, PE
Goat Polyclonal Secondary Antibody
Brand: Invitrogen™ P21129
This item is not returnable.
View return policy
Description
Goat anti-Mouse IgG1 Cross-Adsorbed Secondary Antibody for Flow, ICC/IF
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
| Mouse IgG1 Cross-Adsorbed | |
| Polyclonal | |
| PE | |
| Goat | |
| Purified | |
| RUO | |
| Mouse | |
| Secondary Antibody | |
| IgG |
| Flow Cytometry, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 1% Prionex, 2mM EDTA, 1% glycerol and 5mM sodium azide; pH 7.5 | |
| IgG gamma 1. | |
| 250 μL | |
| Secondary | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction