missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Mouse IgG1 Secondary Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-69525
This item is not returnable.
View return policy
Description
IgG1 Polyclonal antibody specifically detects IgG1 in Mouse samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| IgG1 | |
| Polyclonal | |
| Unconjugated | |
| Whole antiserum | |
| Goat | |
| Whole Antiserum | |
| RUO | |
| Secondary | |
| Mouse | |
| Lyophilized |
| Immunofluorescence | |
| LYOPH | |
| Immunocytochemistry/ Immunofluorescence 1:10 - 1:500 | |
| IgG1, Igh-4, RGD1359539, VH7183 | |
| Pools of highly purified homogenous IgG1 isolated from mouse serum. Freund's complete adjuvant is used in the first step of the immunization procedure. | |
| 1 mL | |
| Immunology | |
| Reconstitute the lyophilized antiserum by adding 1 mL sterile distilled water. Dilutions may be prepared by adding phosphate buffered saline (PBS, pH 7.2). | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction