missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Mouse IgG2a Secondary Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-69153
This item is not returnable.
View return policy
Description
IgG2a Polyclonal antibody specifically detects IgG2a in Mouse samples. It is validated for ELISA, Immunoelectrophoresis
Specifications
| IgG2a | |
| Polyclonal | |
| Unconjugated | |
| Whole antiserum | |
| Pools of purified homogenous IgG2a isolated from mouse serum of strains belonging to the allelic types Igh-1a and Igh-1b. Freund's complete adjuvant is used in the first step of the immunization procedure. | |
| 1 mL | |
| Secondary | |
| Mouse | |
| Lyophilized |
| ELISA, Immunoelectrophoresis | |
| LYOPH | |
| ELISA, Immunoelectrophoresis | |
| Goat | |
| Whole Antiserum | |
| RUO | |
| Reconstitute the lyophilized antiserum by adding 1 mL sterile distilled water. Dilutions may be prepared by adding phosphate buffered saline (PBS, pH 7.2). | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction