Learn More
Goat anti-Mouse IgM Fc Secondary Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand:  Novus Biologicals NBP2-69733
Product Code. 18621758
  • 502.00 € / 1mL
Description
IgM Fc Polyclonal antibody specifically detects IgM Fc in Mouse samples. It is validated for ELISA, Immunoprecipitation, Immunoelectrophoresis
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IgM Fc
Polyclonal
Unconjugated
Whole antiserum
Highly purified homogenous IgM isolated from mouse serum. Freund's complete adjuvant is used in the first step of the immunization procedure.
1 mL
B Cell Development and Differentiation Markers, Immunology
Reconstitute the lyophilized antiserum by adding 1 mL sterile distilled water. Dilutions may be prepared by adding phosphate buffered saline (PBS, pH 7.2).
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
ELISA, Immunoprecipitation, Immunoelectrophoresis
LYOPH
ELISA, Immunoprecipitation 1:10 - 1:500, Immunoelectrophoresis
Goat
Whole Antiserum
RUO
Secondary
Mouse
Lyophilized