missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Rabbit Ig, Clone: Poly1272, Secondary Antibody, BD
Description
X
Host Species: Goat
Clone: Poly1272
Isotype: Ig
Species Reactivity: Rabbit
Immunohistochemistry, Immunohistochemistry (Frozen)
Specifications
Specifications
| Antigen | Goat anti-Rabbit Ig |
| Classification | Polyclonal |
| Clone | POLYCLONAL |
| Conjugate | Biotin |
| Host Species | Goat |
| Quantity | 1 mL |
| Regulatory Status | RUO |
| Research Discipline | Immunology |
| Primary or Secondary | Secondary |
| Target Species | Rabbit |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?