missing translation for 'onlineSavingsMsg'
Learn More

Goat anti-Rabbit Ig, Polyclonal, FITC, Secondary Antibody, BD

Brand:  BD Biosciences 554020


Product Code. 15807548

  • ONLINE EXCLUSIVE PRICE
    256.00 €
    245.99 € / Each
    Save 10.01 €
    3% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

X

Host Species: Goat
Species Reactivity: Rabbit

Flow Cytometry

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Goat anti-Rabbit Ig
POLYCLONAL
FITC
RUO
Secondary
Ig
Polyclonal
0.5mg/mL
0.5 mg
Immunology
Rabbit
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • FITC Goat Anti-Rabbit Ig

Signal Word
  • Warning

Hazard Category
  • Skin sensitiser Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : CMIT/MIT

SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.