missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Goat anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, HRP

Product Code. 15393916
Change view
Click to view available options
Quantity:
1 mg
50 μg
Unit Size:
1mg
50µg
Product Code. Quantity unitSize
15393916 50 μg 50µg
15281388 1 mg 1mg
2 options
This item is not returnable. View return policy

Product Code. 15393916

Brand: Invitrogen™ A16104SAMPLE

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Goat Polyclonal Secondary Antibody

Rehydrate with 1.1 mL of deionized water and let stand 30 minutes at room temperature to dissolve. (Product has been overfilled to ensure complete recovery.) Centrifuge to remove any particulates. Prepare fresh working dilution daily. Store lyophilized material at 2-8 °C. For long term storage after reconstitution, dilute with 50% glycerol and store at -20 °C as a liquid. Based on Immunoelectrophoresis, no reactivity is observed to: non-immunoglobulin rabbit serum proteins, serum proteins from bovine, human, or mouse , IgG from human or mouse. Based on IEP, this antibody reacts with: heavy (gamma) chains on rabbit IgG; light chains on all rabbit immunoglobulins. Affinity purified antibody is > 95% based on SDS-PAGE.

This product is a sample size (50μG) of Goat anti-Rabbit IgG (H+L) - cross-adsorbed, horseradish peroxidase (HRP) Conjugate, (A16104).

Goat Anti-Rabbit IgG (H+L) Antibody, Horseradish Peroxidase (HRP) Conjugate, is prepared from antibodies that have been adsorbed against bovine, human, and mouse IgG to minimize cross reactivity. Goat Anti-Rabbit IgG (H+L) Antibody, Horseradish Peroxidase (HRP) Conjugate, should be suitable for western blot, ELISA, immunohistochemistry and other standard immunoassay applications. The sensitivity of each lot of antibody is confirmed using ELISA. The specificity of each lot of antibody is confirmed by isoelectric focusing (IEF).

Applications: Goat Anti-Rabbit IgG (H+L) Antibody, Horseradish Peroxidase (HRP) Conjugate, should be suitable for western blot, ELISA, immunohistochemistry and other standard immunoassay applications.
Host species: The host species of theanti-rabbit IgG (H+L), horseradish peroxidase (HRP) Conjugate, is goat.
Reactivity: Goat Anti-Rabbit IgG (H+L) Antibody, Horseradish Peroxidase (HRP) Conjugate, detects rabbit IgG (H+L).
Product size: 50μg
Suggested dilution: The optimal working dilution should be determined empirically. Dilutions of 1:500-1:5000 should be satisfactory for most immunohistochemical applications. Dilutions of 1:200-1:5,000 should be satisfactory for most ELISA and western blot applications.

TRUSTED_SUSTAINABILITY

Specifications

Applications Immunoassay, Immunohistochemistry, Western Blot
Conjugate HRP
Description Goat anti-Rabbit IgG (H+L) - Cross-adsorbed, 50μg, Lyophilized, Goat Host, Mouse Species Reactivity, HRP Conjugate, IgG Isotype, 2 to 8°C Storage Temperature, For Cellular Imaging, Protein Assays and Analysis, Secondary Detection
Formulation Lyophilized
Host Species Goat
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Protein Assays and Analysis
Whole Molecule Whole Antibody
Primary or Secondary Secondary
Packaging 1 Each
Target Species Mouse
Content And Storage Store at 2–8°C. For long term storage of antibody solutions, dilute with 50% glycerol, aliquot, and store at -5 to -35°C.
Form Lyophilized
Isotype IgG
Reactivity Mouse
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.