Learn More
Goat anti-Rabbit IgG (H+L) Highly Cross-Adsorbed, HRP, Polyclonal, Secondary Antibody SAMPLE, Invitrogen™
Goat Polyclonal Secondary Antibody
Brand: Invitrogen A16110SAMPLE
Additional Details : Weight : 0.01000kg
Description
Anti-Rabbit secondary antibodies are affinity-purified antibodies with well-characterized specificity for rabbit immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
Rabbit IgG (H+L) Highly Cross Adsorbed | |
Polyclonal | |
PBS with 1% BSA and 0.1% Kathon CG; pH 7.2 | |
Gamma Immunoglobins Heavy and Light chains | |
1 mg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |
ELISA, Immunohistochemistry, Western Blot | |
HRP | |
Goat | |
Affinity chromatography | |
RUO | |
Rabbit | |
Lyophilized |