Learn More
Invitrogen™ Goat anti-Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor™ Plus 405
Description
To minimize cross-reactivity, the goat anti-rabbit IgG whole antibodies have been affinity-purified and cross-adsorbed against human, mouse and rat serum proteins. Cross-adsorption or pre-adsorption is a purification step to increase specificity of the antibody resulting in higher sensitivity and less background staining. The secondary antibody solution is passed through a column matrix containing immobilized serum proteins from potentially cross-reactive species. Only the nonspecific-binding secondary antibodies are captured in the column, and the highly specific secondaries flow through. The benefits of this extra step are apparent in multiplexing/multicolor-staining experiments (e.g., flow cytometry) where there is potential cross-reactivity with other primary antibodies or in tissue/cell fluorescent staining experiments where there may be the presence of endogenous immunoglobulins. Using conjugate solutions: Centrifuge the protein conjugate solution briefly in a microcentrifuge before use; add only the supernatant to the experiment. This step will help eliminate any protein aggregates that may have formed during storage, thereby reducing nonspecific background staining. Because staining protocols vary with application, the appropriate dilution of antibody should be determined empirically. Specificity: This antibody binds to heavy chains on rabbit IgG and light chains on all rabbit immunoglobulins. This antibody does not bind to human, mouse or rat serum proteins/IgG or...
Specifications
Specifications
| Antigen | Rabbit IgG (H+L) Highly Cross-Adsorbed |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 2 mg/mL |
| Conjugate | Alexa Fluor Plus 405 |
| Formulation | Proprietary buffer with 0.016% Methylisothiazolone, 0.016% Bromonitrodioxane; pH 6.5 |
| Host Species | Goat |
| Immunogen | Gamma Immunoglobins Heavy and Light chains. |
| Purification Method | Affinity chromatography |
| Quantity | 1 mg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.