missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Goat anti-Rabbit IgG (H+L) Secondary Antibody, HRP

Product Code. 10035943
Change view
Click to view available options
Quantity:
2 mL
Unit Size:
2mL
Product Code. Quantity unitSize
10035943 2 mL 2mL
1 options
This item is not returnable. View return policy

Product Code. 10035943

Brand: Invitrogen™ 32460

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Goat Polyclonal Secondary Antibody

This antibody may cross-react with immunoglobulins from other species. Working dilutions for Western blotting using SuperSignal West Substrates: SuperSignal West Pico Substrate - 1:250-1:1250 SuperSignal West Dura Substrate - 1:600-1:3000 SuperSignal West Femto Substrate - 1:1250-1:6000 ECL Western Blotting Substrate - 1:20-1:125.

Anti-Rabbit secondary antibodies are affinity-purified antibodies with well-characterized specificity for rabbit immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rabbit IgG (H+L)
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.01 mg/mL
Conjugate HRP
Formulation PBS with proprietary stabilizer and no preservative
Host Species Goat
Immunogen Purified Rabbit IgG, whole molecule.
Purification Method Antigen affinity chromatography
Quantity 2 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Rabbit
Content And Storage 4°C
Product Type Secondary Antibody
Form Liquid
Isotype IgG
Show More Show Less
compliance-icons
Product Identifier
  • GOAT ANTI-RABBIT CONJUGATED
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Mixture, 3(2H)-isothiazolone, 5-chloro-2-methyl- with 2-methyl-3(2H)-isothiazolone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.