missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Rabbit IgG (H+L) Secondary Antibody (Pre-adsorbed), Novus Biologicals™
Goat Polyclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NBP1-75321-0.5mg
This item is not returnable.
View return policy
Description
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Rabbit samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunodiffusion, Immunoassay.
Specifications
| IgG (H+L) | |
| Polyclonal | |
| Unconjugated | |
| Purified Rabbit IgG (H&L) | |
| 0.5 mg | |
| Adaptive Immunity, Immunology | |
| Based on IEP, this antibody reacts with heavy gamma chains on rabbit IgG and light chains on all rabbit immunoglobulins. This antibody has been pre-adsorbed against bovine, goat, human, mouse and rat IgG and serum. | |
| Store at 4°C. After dilution, add an equal volume of glycerol to prevent loss of enzymatic activity and store at -20°C. Avoid repeated freeze/thaw cycles. |
| Immunoassay | |
| 4.5 mg/mL | |
| Goat | |
| Affinity purified | |
| RUO | |
| Secondary | |
| Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction