Learn More
Invitrogen™ Goat anti-Rabbit IgG (Heavy chain) Superclonal™ Secondary Antibody, Alexa Fluor™ 488
Goat Recombinant Superclonal Secondary Antibody
Brand:  Invitrogen™ A27034SAMPLE
 View more versions of this product
Product Code. 15677868
  • 80.75 € / 0.05mg
Description
The sensitivity and specificity of each lot is confirmed using ELISA. Minimal cross-reactivity with mouse, rat, human, bovine, guinea pig and donkey IgG is observed. Recombinant antibodies are produced using specific genes that code for the desired antibodies. These genes are cloned into an expression vector and expressed in vitro. The advantages of recombinant antibodies include: better specificity, animal origin-free formulation, and more lot-to-lot consistency.
Anti-Rabbit secondary antibodies are affinity-purified antibodies with well-characterized specificity for rabbit immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Rabbit IgG (Heavy chain)
Recombinant Superclonal
Alexa Fluor 488
Goat
Gravity column chromatography
RUO
Rabbit
Secondary Antibody
IgG
Flow Cytometry, Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry
1 mg/mL
PBS with 5mM sodium azide
Recombinant full-length protein.
50 μg
Secondary
4°C, store in dark, DO NOT FREEZE!