missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Rat IgG2a/IgG2b Secondary Antibody, FITC, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-69589
This item is not returnable.
View return policy
Description
IgG2a/IgG2b Polyclonal antibody specifically detects IgG2a/IgG2b in Rat samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Frozen)
Specifications
| IgG2a/IgG2b | |
| Polyclonal | |
| FITC | |
| Lyophilized from PBS (pH 7.2) | |
| Pools of purified homogenous IgG2a and IgG2b isolated from rat serum. Freund's complete adjuvant is used in the first step of the immunization procedure. | |
| 1 mL | |
| Secondary | |
| Rat | |
| Lyophilized |
| ELISA, Immunofluorescence, Immunohistochemistry (Frozen) | |
| LYOPH | |
| ELISA 1:100-1:2000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Frozen | |
| Goat | |
| Ion exchange chromatography | |
| RUO | |
| Reconstitute by by adding 1 mL sterile distilled water, spun down to remove insoluble particles, divided into small aliquots, frozen and stored at or below -20C. | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction