missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat IgG Fraction to Human Kappa Light Chain (free), FITC, Polyclonal Secondary Antibody, MP Biomedicals
Brand: MP Biomedicals 0855159
This item is not returnable.
View return policy
Description
Host Species: Goat
Species Reactivity: Hu
Immunofluorescence, Fluorescence Microscopy, Fluorescent Activated Cell Sorting, Western Blotting
Specifications
| Kappa Light Chain | |
| FITC | |
| 2 mL | |
| Secondary | |
| Purified |
| Polyclonal | |
| Goat | |
| RUO | |
| Human |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction