missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Goat Serum

Product Code. 11475055 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11475055 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11475055 Supplier MP Biomedicals™ Supplier No. 092939149
  • ONLINE EXCLUSIVE PRICE
    35.45 €
    33.60 € / 100mL
    Save 1.85 €
    5% Off
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Suitable alternative to fetal bovine serum in a wide range of cell culture applications such as the isolation and identification of viruses produced in a variety of primary cell lines.

This serum is produced from whole blood collected from healthy adult animals.

Specifications

Color Reddish brown
Content And Storage Store at -20°C
Form Liquid
Quantity 100 mL
Sterilization Method 0.2 μm filtration
Source Whole blood collected from healthy adult animals
For Use With (Application) Cell Culture
pH 7 to 8
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.