Learn More
3M™ GoggleGear™ Protective Goggle 3000 Series, PVC Shroud, Clear AF Lens, Premium Cloth Strap
Product Code. 31875401 Shop All 3M Products
Change view
Click to view available options
Frame Material:
PVC
Unit Size:
Each
Product Code. Frame Material Unit Size
31875401 PVC Each
1 options
Product Code. 31875401 Supplier 3M™ Supplier No. 7100325039
  • ONLINE EXCLUSIVE PRICE
    20.65 €
    13.08 € / Pack of 1
    Save 7.57 €
    36% Off
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Protective goggle 3000 series features a full-size design with wraparound lens for improved field of vision, and slotted nose bridge for a more comfortable fit
  • 3M™ GoggleGear™ Protective Goggle 3000 Series features a full-size design with wrap-around lens for improved field of vision, and slotted nose bridge for a more comfortable fit.
  • Choose from models with anti-fog or 3M™ Scotchgard™ Anti-Fog coating.
  • The GoggleGear™ 3000 is available with indirect venting for enhanced air circulation and anti-fog performance, and is 3, protection against droplets, 4, protection against large particles, and 6, protection against streams of liquid, rated.
  • Includes either an adjustable polyester cloth strap, or easy to clean adjustable neoprene strap, as well as an optimized shroud design with slots to accommodate many prescription eyewear.
  • Acetate lens model meets the chemical resistance requirement of ISO 16321, helping protect workers from chemical hazards.
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog
Color Clear
Lens Tint Clear
Product Type Anti-fog Goggles
Disposable Reusable
Lens Material Acetate
Frame Material PVC
UV Protection None
Frame Type Full Frame
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.