missing translation for 'onlineSavingsMsg'
Learn More

Gold Germanium slug, 3.175mm (0.125 in.) dia. x≈3.175mm (0.125 in.) length, Premion™, 99.998% (metals basis)

Product Code. 15413885
Change view
Click to view available options
Quantity:
25 Pc.
5 Pc.
Unit Size:
Pack of 25
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15413885 5 Pc. Pack of 5
15423885 25 Pc. Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15413885 Supplier Thermo Scientific Alfa Aesar Supplier No. 042983.KJ
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

Gold Germanium slug is used in scientific research and development activities.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Gold Germanium slug
Name Note 3.175mm (0.125 in.) dia. x≈3.175mm (0.125 in.) length
Avg. Mol. Wt. or Mol. Wt. Range Au:Ge; 88:12 wt%
Density 0.37 g/cm3
Quantity 5 Pc.
Odor Odorless
Weight ≈0.4g/slug
Form Slug
Assay Percent Notes (metals basis)
MDL Number MFCD02091706
Solubility Information Insoluble in water.
Percent Purity 99.998%
Grade Premion
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.