missing translation for 'onlineSavingsMsg'
Learn More

Gold(I) iodide, 99%

Product Code. 11397188
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11397188 5 g 5g
11307198 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 11397188

Brand: Thermo Scientific Alfa Aesar 016617.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10294-31-2 | AuI | 323.87 g/mol

Gold(I) iodide is employed in the synthesis of chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10294-31-2
Molecular Formula AuI
Molecular Weight (g/mol) 323.87
MDL Number MFCD00014175
InChI Key ATGIETUGWDAYPU-UHFFFAOYSA-M
Synonym gold monoiodide, gold i iodide, gold iodide, acmc-1bqvl, gold i iodide trace metals basis
PubChem CID 82526
IUPAC Name iodogold
SMILES [I-].[Au+]

Specifications

Melting Point 120°C (decomposition)
Physical Form Crystalline
Quantity 5 g
UN Number UN3260
Solubility Information Insoluble in water; Soluble in alkali iodide, cyanide solutions; Decomposes in warm acids
Formula Weight 323.87
Percent Purity 99%
Sensitivity Moisture sensitive
Density 8.25 g/mL
Chemical Name or Material Gold(I) iodide
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Gold iodide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.