missing translation for 'onlineSavingsMsg'
Learn More

Gold nanoparticles, 20nm, supplied in 0.1mM PBS, 95%, OD1, 524 nm absorption

Product Code. 15460727
Change view
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15460727 25 mL 25mL
15494585 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15460727 Supplier Thermo Scientific Alfa Aesar Supplier No. J67318.AC
 Restricted Item
Request Bulk or Custom Format

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

CAS: 7440-57-5 | Au | 196.97 g/mol

For development of peptide and protein gold conjugates for use in applications such as blotting, microscopy and TEM

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • For development of peptide and protein gold conjugates for use in applications such as blotting, microscopy and TEM
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-57-5
Molecular Formula Au
Molecular Weight (g/mol) 196.97
MDL Number MFCD00003436
InChI Key PCHJSUWPFVWCPO-UHFFFAOYSA-N
Synonym colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
PubChem CID 23985
ChEBI CHEBI:30050
IUPAC Name gold
SMILES [Au]

Specifications

Color Red
Quantity 25 mL
Particle Size 20nm
Sensitivity Light sensitive
Form Liquid
Percent Purity 95%
Chemical Name or Material Gold nanoparticles, supplied in 0.1mM PBS, 95%, OD: 1, 524 nm absorption
I am using Gold nanoparticles, 20 nm, supplied in 0.1 mM PBS, 95%, OD1, 524 nm absorption (Cat. No. J67318.xx). How do I ensure my gold nanoparticle conjugation reaction was successful?

We recommend using immunoblotting (dot-blot) to test a gold conjugate for the presence of the conjugated antibody (or antigen). Another alternative is to directly use the gold conjugate in the downstream application and compare its performance to a negative control gold conjugate.

I have the Gold nanoparticles, 20 nm, supplied in 0.1 mM PBS, 95%, OD1, 524 nm absorption (Cat. No. J67318.xx). Why does the gold nanoparticle solution turn violet when I add salt-containing buffer?

The normal color of the gold nanoparticles is red. Due to repulsive forces arising from the surface charge of gold nanoparticles, an energy barrier must be overcome for individual particles to interact. When no, or a small, amount of electrolytes such as NaCl are present, this energy barrier is too strong for interaction to occur between particles. However, once NaCl is added, this energy barrier is reduced and allows the gold nanoparticles to interact and aggregate. This aggregation causes a phenomenon called "surface-plasmon coupling" which changes the adsorption maximum of light to a higher wavelength, resulting in a change in color of the solution.

At what temperature should I store my Gold nanoparticles, 20 nm, supplied in 0.1 mM PBS, 95%, OD1, 524 nm absorption (Cat. No. J67318.xx)?

Gold nanoparticles should be stored at 4-25° Celsius. Do not store gold nanoparticles in the freezer since this cause irreversible aggregation.

My Gold nanoparticles, 20 nm, supplied in 0.1 mM PBS, 95%, OD1, 524 nm absorption (Cat. No. J67318.xx) settles out of solution upon storage. Is this normal?

Settling of gold nanoparticles on the bottom of the bottle is normal and is especially common for larger sized particles, which settle at a greater speed. Settling does not affect the performance of the particles. Prior to use, we recommend swirling the solution to properly disperse your gold nanoparticles.

How do I wash my Gold nanoparticles, 20 nm, supplied in 0.1 mM PBS, 95%, OD1, 524 nm absorption (Cat. No. J67318.xx)?

For most applications, our gold nanoparticles can be used without any additional washing steps. If you have a sensitive application that requires additional washing, we recommend centrifuging the gold nanoparticles. The easiest way to remove possible contaminants in the nanoparticle solution is by centrifugation. We recommend the following protocol:

  1. Place 1 mL aliquot of colloidal gold in a 1.5 mL micro centrifuge tube.
  2. Centrifuge the gold nanoparticles for 30 minutes at 10,000 RPM.
  3. Remove the supernatant and re-suspend in an appropriate volume of ultra-pure water.
  4. Vortex to re-disperse the particles.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.